Nisin

Supplier: Thermo Scientific Chemicals

J66370-03 J66370-06 J66370-14
10067-404EA 63.08 USD
10067-404 10067-406 10067-408
Nisin
Nisin

Effective against a broad range of Gram-positive bacteria but not against Gram-negative cell membranes


  • 900 I.U./mg


CAS No.: 1414-45-5 Molecular Formula: C143H230N42O37S7 Formula Weight: 3354.06 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: Pale brown Boiling Point: 2967° Solubility: Soluble in water MDL No.: MFCD00131724


Applications: Effective against a broad range of gram positive bacteria but not against gram negative cell membranes

Formula: C₁₄₃H₂₃₀N₄₂O₃₇S₇
MW: 3354.06 g/mol
Boiling Pt: 2967 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00131724
CAS Number: 1414-45-5
EINECS: 215-807-5
Merck Index: 14,6564

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR