Mupirocin 92%
Supplier: Thermo Scientific Chemicals
Synonyms:
9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid
J66347-MC
J66347-MF
10067-454EA
800
USD
10067-454
10067-456
Mupirocin 92%
Mupirocin
Mupirocin, 92-102%
Purity: 92%
CAS No.: 12650-69-0
Molecular Formula: C26H44O9
Formula Weight: 500.62
Flash Point:
Formula:
C₂₆H₄₄O₉ MW: 500.629 g/mol Boiling Pt: 672 °C Melting Pt: 77…78 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01711620 CAS Number: 12650-69-0 Merck Index: 14,6302 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...