Netilmicin sulfate 595 µg/mg
Supplier: Thermo Scientific Chemicals
|
Danger
|
A broad spectrum antibiotic
- Light Sensitive
- Hygroscopic
White to off-white powder
This antibiotic is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied.
Soluble in water
Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₂₁H₄₁N₅O₇·⁵/₂H₂SO₄ Boiling Pt: 868 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01311715 CAS Number: 56391-57-2 EINECS: 260-147-3 Merck Index: 14,6479 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...