Puromycin aminonucleoside 98%

Supplier: Thermo Scientific Chemicals
Warning

J66301-MC J66301-ME
10067-018EA 533 USD
10067-018 10067-020
Puromycin aminonucleoside 98%
Puromycin aminonucleoside

A puromycin aminonucleoside useful in the study of human glomerular disease.


White powder


Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.


Soluble in water at 50 mg/mL. May require heating


Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₁₂H₁₈N₆O₃
MW: 294.31 g/mol
Melting Pt: 215…216 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063462
CAS Number: 58-60-6
EINECS: 200-388-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR