Cefepime hydrochloride monohydrate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cefepime Dihydrochloride Monohydrate, 1-(((6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)-1-methylpyrrolidin-1-ium chloride hydrochloride hydrate
A broad spectrum antibiotic targeting a wide variety of naturally antibiotic resistant Gram-positive and Gram-negative bacteria
- Light sensitive
- White to pale yellow powder
CAS No.: 123171-59-5
Molecular Formula: C19H24N6O5S2.HCl.H2O
Formula Weight: 571.5
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to pale yellow
Melting Point: 150°
Solubility: Soluble in DMSO
MDL No.: MFCD01687679
Applications: A broad spectrum antibiotic targeting a wide variety of naturally antibiotic resistant gram positive and gram negative bacteria
Soluble in DMSO
Formula:
C₁₉H₂₅ClN₆O₅S₂·HCl·H₂O Melting Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD01687679 CAS Number: 123171-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...