Sulbactam 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sulbactam acid, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 4,4-Dioxide

J66235-03 J66235-06
10067-100EA 94.66 USD
10067-100 10067-102
Sulbactam 98%
Sulbactam

Commonly used beta-lactam antibiotics to prevent degradation by beta-lactamase enzymes


Applications: Commonly used beta-lactam antibiotics to prevent degradation by beta-lactamase enzymes


Applications: Commonly used beta-lactam antibiotics to prevent degradation by beta-lactamase enzymes

Formula: C₈H₁₁NO₅S
MW: 233.24 g/mol
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00867005
CAS Number: 68373-14-8
EINECS: 269-878-2
Merck Index: 14,8889

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR