Tazobactam sodium

Supplier: Thermo Scientific Chemicals

J66226-MC J66226-03
10067-012EA 80.81 USD
10067-012 10067-098
Tazobactam sodium
Tazobactam sodium

Tazobactam sodium


CAS No.: 89785-84-2 Molecular Formula: C10H11N4NaO5S Formula Weight: 322.27 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 140-147° Boiling Point: 77° Solubility: Soluble in water MDL No.: MFCD00917472

Formula: C₁₀H₁₁N₄NaO₅S
Boiling Pt: 77 °C
Melting Pt: 140…147 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00917472
CAS Number: 89785-84-2
Merck Index: 14,9083

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR