Roxithromycin 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3R,4S,5S,6R,7R,9R,11S,12R,13S,14R,E)-6-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-10-(((2-methoxyethoxy)methoxy)imino)-3,5,7,9,11,13-hexamethyloxacyclotetradecan-2-one
Roxithromycin is used to block bacterial protein synthesis
- White powder
Clinically, used to treat respiratory tract, urinary and soft tissue infections.
Soluble in water to 18 μg/mL
Precaución: Research use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₄₁H₇₆N₂O₁₅ MW: 837.04 g/mol Melting Pt: 111…118 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00214389 CAS Number: 80214-83-1 Merck Index: 14,8276 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...