Praziquantel 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one

J66188-06 J66188-14
10067-072EA 77.4 USD
10067-072 10067-074
Praziquantel 98%
Praziquantel

An anti-helminthic compound


Applications: An anti-helminthic compound


Applications: An anti-helminthic compound

Formula: C₁₉H₂₄N₂O₂
MW: 312.41 g/mol
Boiling Pt: 1377 °C
Melting Pt: 136…138 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00058531
CAS Number: 55268-74-1
EINECS: 259-559-6
Merck Index: 14,7716

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR