Glufosinate-ammonium 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
DL-Phosphinothricin, monoammonium salt
J66186-MD
J66186-03
10067-008EA
178.91
USD
10067-008
10067-070
Glufosinate-ammonium 95%
Glufosinate ammonium
Interferes with the synthesis of the amino acid glutamine
Purity: 95%
CAS No.: 77182-82-2
Molecular Formula: C5H15N2O4P
Formula Weight: 198.16
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: Interferes with the synthesis of the amino acid glutamine
Formula:
C₅H₁₈N₃O₄P Boiling Pt: 519 °C Melting Pt: 215 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00055562 CAS Number: 77182-82-2 EINECS: 278-636-5 Merck Index: 14,7339 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...