Ribostamycin sulphate 630 µg/mg

Supplier: Thermo Scientific Chemicals
Danger

J66155-MD J66155-03
10067-006EA 256 USD
10067-006 10067-068
Ribostamycin sulphate 630 µg/mg
Ribostamycin sulphate

Ribostamycin sulfate is an broad spectrum antibiotic used in pharmacokinetic studies.


White to pale yellow powder


Applications: A broad spectrum antibiotic targeting both gram positive and gram negative bacteria


.


Soluble in water


Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₁₇H₃₄N₄O₁₀·H₂SO₄
Melting Pt: 175…180 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00057496
CAS Number: 53797-35-6
EINECS: 258-783-1
Merck Index: 14,8206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR