Piperacillin 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(2S,5R,6R)-6-((R)-2-(4-Ethyl-2,3-Dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
J66143-03
J66143-ME
10067-062EA
195
USD
10067-062
AAJ66143-ME
Piperacillin 95%
Piperacillin
Piperacillin, 95%
Purity: 95%
CAS No.: 61477-96-1
Molecular Formula: C23H27N5O7S
Formula Weight: 517.55
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 139-140°
Solubility: Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL
MDL No.: MFCD00865043
Formula:
C₂₃H₂₇N₅O₇S MW: 517.557 g/mol Melting Pt: 139…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00865043 CAS Number: 61477-96-1 EINECS: 262-811-8 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...