Brij® 35, solid for electrophoresis
Supplier: Thermo Scientific Chemicals
Synonyms:
Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
J66096-0B
J66096-30
AAJ66096-0BEA
61
USD
AAJ66096-0B
AAJ66096-30
Brij® 35, solid for electrophoresis
Brij® 35
Brij® L23 Electrophoresis Reagent
CAS No.: 9002-92-0
Molecular Formula: C58H11O24
Formula Weight: 1199.57
Physical Form: Waxy solid
Appearance: White
Melting Point: ca 35-40°
Solubility: Soluble in water
MDL No.: MFCD00080891
Formula:
C₅₈H₁₁₈O₂₄ MW: 1199.56 g/mol Melting Pt: ∼ 35…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00080891 CAS Number: 9002-92-0 EINECS: 500-002-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...