Cefixime trihydrate
Supplier: Thermo Scientific Chemicals
|
Danger
|
J66094-MC
J66094-03
10067-004EA
380.42
USD
10067-004
10067-056
Cefixime trihydrate
Cefixime trihydrate
A broad spectrum third generation cephalosporin
- Air and Light Sensitive
CAS No.: 125110-14-7
Molecular Formula: C16H15N5O7S2.3H2O
Formula Weight: 507.49
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Light Sensitive
Appearance: Pale yellow
Melting Point: 218-225°
Solubility: Soluble in water at 55.1 µg/mL
MDL No.: MFCD03788802
Applications: A broad spectrum third generation cephalosporin
Formula:
C₁₆H₂₁N₅O₁₀S₂ Melting Pt: 218…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788802 CAS Number: 125110-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...