Sisomicin sulphate Salt 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis((2R,3R,4R,5R)-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-5-methyl-4-(methylamino)tetrahydro-2H-pyran-3,5-diol) pentakis(sulphate), Sisomicin sulphate

J66063-03 J66063-ME
10067-076EA 292 USD
10067-076 AAJ66063-ME
Sisomicin sulphate Salt 90%
Sisomicin sulphate Salt

Sisomicin is a broad spectrum aminoglycoside antibiotic that targets the 30S ribosomal subunit resulting in an inability to read mRNA


  • White to off-white powder


Sisomicin is related to gentamicin, but is more effective against Pseudomonas aeruginosa.


Soluble in water

Formula: (C₁₉H₃₇N₅O₇)₂·5H₂SO₄
MW: 1385.4 g/mol
Boiling Pt: 155 °C
Melting Pt: 70…73 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00058308
CAS Number: 53179-09-2
EINECS: 258-414-4
Merck Index: 14,8550

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR