all cis-5,8,11,14,17-Eicosapentaenoic Acid
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic Acid, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, EPA
J66060-MC
J66060-ME
AAJ66060-MCEA
221
USD
AAJ66060-MC
AAJ66060-ME
all cis-5,8,11,14,17-Eicosapentaenoic Acid
all cis-5,8,11,14,17-Eicosapentaenoic acid
cis-5,8,11,14,17-Eicosapentaenoic acid
CAS No.: 10417-94-4
Molecular Formula: C20H30O2
Formula Weight: 302.45
Storage Temperature: -30°C to -10°C
Flash Point: 93°(199°F)
Physical Form: Oil
Density: 0.943
Refractive Index: 1.4975
Sensitivity: Air and Light Sensitive
Appearance: Colorless
Melting Point: -54°
Solubility: Soluble in methanol
Beilstein Reference: 1714433
UN No.: 3265
MDL No.: MFCD00065716
Formula:
C₂₀H₃₀O₂ MW: 302.46 g/mol Melting Pt: –54 °C Density: 0.943 Flash Pt: 93 °C (199 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065716 CAS Number: 10417-94-4 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...