Midecamycin 900 µg/mg

Supplier: Thermo Scientific Chemicals

Synonyms: (4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-(((2S,3R,4R,5S,6R)-4-(Dimethylamino)-3-hydroxy-5-(((2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-(propionyloxy)tetrahydro-2H-pyran-2-yl)oxy)-6-methyltetrahydro-2H-pyran-2-yl)oxy)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)oxacyclohexadeca-11,13-dien-4-yl propionate

J66046-ME J66046-03
10067-002EA 171 USD
10067-002 10067-048
Midecamycin 900 µg/mg
Midecamycin

A broad spectrum antibiotic


White powder


Applications: A broad spectrum antibiotic


Soluble in methanol or ethanol. Insoluble in water

Formula: C₄₁H₆₇NO₁₅
MW: 813.97 g/mol
Boiling Pt: 874 °C
Melting Pt: 155…156 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00865000
CAS Number: 35457-80-8
EINECS: 252-578-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR