Gatifloxacin sesquihydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid sesquihydrate

J66036-03 J66036-06
10066-678EA 36.4 USD
10066-678 10066-990 10066-992
Gatifloxacin sesquihydrate
Gatifloxacin sesquihydrate

A broad spectrum antibiotic


CAS No.: 180200-66-2 Molecular Formula: C19H22FN3O4.1.5H2O Formula Weight: 402.42 Physical Form: Powder Appearance: White to pale yellow Boiling Point: 684° Solubility: Soluble in 1N NaOH at 10mg/ml MDL No.: MFCD22373645


Applications: A broad spectrum antibiotic

Formula: C19H22FN3O4•1.5H2O
MW: 804.83 g/mol
Boiling Pt: 684 °C
Storage Temperature: Ambient
MDL Number: MFCD22373645
CAS Number: 180200-66-2
Merck Index: 14,4376

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR