Spectinomycin sulphate 93%

Supplier: Thermo Scientific Chemicals
Danger

J66034-03 J66034-06
10066-622EA 48.53 USD
10066-622 10066-624
Spectinomycin sulphate 93%
Spectinomycin sulphate

This spectinomycin sulfate antibiotic acts against Gram-negative bacteria.


White powder


Applications: Targets gram negative bacteria


Soluble in DMSO or water

Formula: C₁₄H₂₄N₂O₇·H₂SO₄
Boiling Pt: 583 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00270185
CAS Number: 23312-56-3
EINECS: 245-578-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR