Clarithromycin 960 - 1040 µg/mg ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-12,13-dihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-7-methoxy-3,5,7,9,11,13-hexamethyloxacyclotetradecane-2,10-dione

J66005-03 J66005-06 J66005-MC
10066-618EA 103.99 USD
10066-618 10066-620 10067-000
Clarithromycin 960 - 1040 µg/mg ≥97%
Clarithromycin

This clarithromycin is a macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit.


  • Optical rotation: ‒92° (c=1 in dichloromethane)


White powder


Optical rotation: ‒92° (c=1 in dichloromethane)


Soluble in DMSO


Precaución: For research use only. Not intended for animal or human diagnostic or therapeutic use.

Formula: C₃₈H₆₉NO₁₃
MW: 747.96 g/mol
Boiling Pt: 86 °C
Melting Pt: 217…220 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00865140
CAS Number: 81103-11-9
Merck Index: 14,2339

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR