Ceftriaxone disodium salt hemiheptahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ceftriaxone sodium hydrate, Ceftriaxone sodium trihydrate, Ceftriaxone sodium salt hydrate, (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-3-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, disodium salt, hemiheptahydrate
Ceftriaxone sodium hemiheptahydrate, 850-995 microg/mg
CAS No.: 104376-79-6
Molecular Formula: C18H16N8Na2O7S3.3.5H2O
Formula Weight: 661.60
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Freely soluble in water
MDL No.: MFCD01750405
Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₁₈H₁₈N₈O₇S₃ MW: 598.53 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01750405 CAS Number: 104376-79-6 EINECS: 277-930-0 Merck Index: 14,1953 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...