Azadibenzocyclooctyne-PEG4-NHS ester

Supplier: Thermo Scientific Chemicals

J64126-MA J64126-MB J64126-MC
10067-572EA 152 USD
10067-572 10067-574 10067-576
Azadibenzocyclooctyne-PEG4-NHS ester
Azadibenzocyclooctyne-PEG4-NHS ester

Azadibenzocyclooctyne-PEG4-NHS ester (ADIBO-PEG4-NHS ester) is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.


  • Form: Oil
  • Storage and sensitivity: Store at −20 °C and moisture sensitive


Precaución: Incompatible with oxidizing agents and heat.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR