3-Aminopicolinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Amino-2-pyridinecarboxylic acid, 3-Aminopyridine-2-carboxylic acid, 2-Pyridinecarboxylic acid, 3-amino-
H64861-06
H64861-14
AAH64861-06EA
68
USD
AAH64861-06
AAH64861-14
3-Aminopicolinic acid 97%
3-Aminopicolinic acid
3-Aminopyridine-2-carboxylic acid, 98%
Purity: 98%
CAS No.: 1462-86-8
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Brown
Melting Point: 218-222°
Beilstein Reference: 3600
MDL No.: MFCD00090153
Formula:
C₆H₆N₂O₂ Melting Pt: 218…222 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00090153 CAS Number: 1462-86-8 EINECS: 215-971-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...