3-Fluorophenethylamine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(3-Fluorophenyl)ethylamine
H64762-06
H64762-14
AAH64762-06EA
39.7
USD
AAH64762-06
AAH64762-14
AAH64762-22
3-Fluorophenethylamine 97+%
3-Fluorophenethylamine
2-(3-Fluorophenyl)ethylamine, 97+%
Purity: 97+%
CAS No.: 404-70-6
Molecular Formula: C8H10FN
Formula Weight: 139.17
Flash Point: 83°(181°F)
Physical Form: Liquid
Density: 1.066
Refractive Index: 1.5090
Boiling Point: 87°/15mm
Beilstein Reference: 3045789
UN No.: 2735
MDL No.: MFCD00075376
Formula:
C₈H₁₀FN MW: 139.17 g/mol Boiling Pt: 87 °C/15 mm Density: 1.066 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075376 CAS Number: 404-70-6 UN: 2735 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...