3-Bromocarbazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-9H-carbazole

H64437-03 H64437-06 H64437-14
AAH64437-03EA 32 USD
AAH64437-03 AAH64437-06 AAH64437-14
3-Bromocarbazole 98%
3-Bromocarbazole

3-Bromocarbazole, 98%


Purity: 98% CAS No.: 1592-95-6 Molecular Formula: C12H8BrN Formula Weight: 246.11 Physical Form: Solid Appearance: White Melting Point: 195-199° Boiling Point: 160°/6mm Beilstein Reference: 142195 MDL No.: MFCD00222621

Formula: C₁₂H₈BrN
MW: 246.11 g/mol
Boiling Pt: 160 °C/6 mm
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00222621
CAS Number: 1592-95-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR