2-methylpyrimidine-5-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64375-03
H64375-06
AAH64375-03EA
80.4
USD
AAH64375-03
AAH64375-06
AAH64375-MD
2-methylpyrimidine-5-carboxylic acid 97%
2-methylpyrimidine-5-carboxylic acid
2-Methylpyrimidine-5-carboxylic acid, 97%
Purity: 97%
CAS No.: 5194-32-1
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 203-205°
Beilstein Reference: 508773
MDL No.: MFCD08235210
Formula:
C₆H₆N₂O₂ Melting Pt: 203…205 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08235210 CAS Number: 5194-32-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...