5-Hydroxypicolinic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-hydroxypyridine-2-carboxylic acid

H64265-03 H64265-06 H64265-14
AAH64265-03EA 32.8 USD
AAH64265-03 AAH64265-06 AAH64265-14
5-Hydroxypicolinic acid 98+%
5-Hydroxypicolinic acid

5-Hydroxypyridine-2-carboxylic acid, 98+%


Purity: 98+% CAS No.: 15069-92-8 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Physical Form: Solid Melting Point: 260-264° Beilstein Reference: 116009 MDL No.: MFCD00661309

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00661309
CAS Number: 15069-92-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR