2-Chloro-4-Bromo-5-fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-4-bromo-5-fluorobenzaldehyde, 4-Bromo-2-chloro-5-fluorobenzaldehyde
H64238-03
H64238-06
AAH64238-03EA
59.1
USD
AAH64238-03
AAH64238-06
AAH64238-MD
2-Chloro-4-Bromo-5-fluorobenzaldehyde 98%
2-Chloro-4-Bromo-5-fluorobenzaldehyde
2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%
Purity: 98%
CAS No.: 1214386-29-4
Molecular Formula: C7H3BrClFO
Formula Weight: 237.45
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD13185877
Formula:
C₇H₃BrClFO MW: 237.452 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185877 CAS Number: 1214386-29-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...