7-Bromo-1-heptene 97%

Supplier: Thermo Scientific Chemicals
Warning

H64127-06 H64127-14 H64127-22
AAH64127-06EA 60.5 USD
AAH64127-06 AAH64127-14 AAH64127-22
7-Bromo-1-heptene 97%
7-Bromo-1-heptene

7-Bromo-1-heptene, 97%


Purity: 97% CAS No.: 4117-09-3 Molecular Formula: C7H13Br Formula Weight: 177.08 Flash Point: 58°(136°F) Physical Form: Liquid Density: 1.162 Refractive Index: 1.4660 Appearance: Colorless to pale yellow Boiling Point: 27°/1mm Beilstein Reference: 1740861 UN No.: 1993 MDL No.: MFCD01631137

Formula: C₇H₁₃Br
MW: 177.085 g/mol
Boiling Pt: 27 °C/1 mm
Density: 1.162
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631137
CAS Number: 4117-09-3
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR