4,4',4''-Triaminotriphenylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(4-aminophenyl)amine
H63963-03
H63963-06
H63963-MD
AAH63963-03EA
102.5
USD
AAH63963-03
AAH63963-06
AAH63963-MD
4,4',4''-Triaminotriphenylamine 97%
4,4',4''-Triaminotriphenylamine
Tris(4-aminophenyl)amine, 97%
Purity: 97%
CAS No.: 5981-09-9
Molecular Formula: C18H18N4
Formula Weight: 290.38
Physical Form: Solid
Melting Point: 230°
Beilstein Reference: 2812659
MDL No.: MFCD00778301
Formula:
C₁₈H₁₈N₄ MW: 290.38 g/mol Melting Pt: 230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00778301 CAS Number: 5981-09-9 EINECS: 227-791-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...