Ethylenedioxybis(trimethylsilane) 98% (by GC)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether), 1,2-Bis(trimethylsiloxy)ethane
H63857-09
H63857-18
H63857-30
AAH63857-09EA
48.3
USD
AAH63857-09
AAH63857-18
AAH63857-30
Ethylenedioxybis(trimethylsilane) 98% (by GC)
Ethylenedioxybis(trimethylsilane)
1,2-Bis(trimethylsiloxy)ethane, 98%
Purity: 98%
CAS No.: 7381-30-8
Molecular Formula: C8H22O2Si2
Formula Weight: 206.43
Flash Point: 51°(124°F)
Refractive Index: 1.4030
Boiling Point: 165-171°
Beilstein Reference: 1744217
UN No.: 1993
MDL No.: MFCD00009640
Formula:
C₈H₂₂O₂Si₂ MW: 206.43 g/mol Boiling Pt: 165…171 °C Density: 0.842 Flash Pt: 51 °C(124 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009640 CAS Number: 7381-30-8 EINECS: 230-950-3 UN: 1993 ADR: "3",III |
Specification Test Results
Assay (GC) | >97.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...