L-Proline-tert-butyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-tert-Butyl pyrrolidine-2-carboxylate, H-Pro-OtBu

H63657-03 H63657-06
AAH63657-03EA 46.6 USD
AAH63657-03 AAH63657-06
L-Proline-tert-butyl ester 98%
L-Proline-tert-butyl ester

L-Proline tert-butyl ester, 98%


Purity: 98% CAS No.: 2812-46-6 Molecular Formula: C9H17NO2 Formula Weight: 171.24 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Colorless MDL No.: MFCD00037879

Formula: C9H17NO2
MW: 171.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037879
CAS Number: 2812-46-6
EINECS: 220-558-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR