2,6-Dichloronicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichloropyridine-3-carbonitrile, 2,6-Dichloro-3-cyanopyridine

H63480-03 H63480-06
AAH63480-03EA 92 USD
AAH63480-03 AAH63480-06 AAH63480-MD
2,6-Dichloronicotinonitrile 97%
2,6-Dichloronicotinonitrile

2,6-Dichloro-3-cyanopyridine, 97%


CAS: 40381-90-6 MDL: MFCD03411341

Formula: C₆H₂Cl₂N₂
MW: 173.00 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD03411341
CAS Number: 40381-90-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR