1-Benzyl-N-carbobenzoxy-L-glutamate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Cbz-L-glutamic Acid 1-Benzyl Ester, N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, Z-Glu-OBzl, 1-Benzyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester
H62975-06
H62975-14
AAH62975-06EA
49.8
USD
AAH62975-06
AAH62975-14
1-Benzyl-N-carbobenzoxy-L-glutamate 95%
1-Benzyl-N-carbobenzoxy-L-glutamate
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%
Purity: 95%
CAS No.: 3705-42-8
Molecular Formula: C20H21NO6
Formula Weight: 371.38
Melting Point: 98-102°
MDL No.: MFCD00077013
Formula:
C₂₀H₂₁NO₆ MW: 371.39 g/mol Melting Pt: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077013 CAS Number: 3705-42-8 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...