Nɑ-(tert-Butoxycarbonyl)-D-lysine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nɑ-Boc-D-lysine, Boc-D-Lys-OH
H62966-03
H62966-06
AAH62966-03EA
67.8
USD
AAH62966-03
AAH62966-06
AAH62966-MD
Nɑ-(tert-Butoxycarbonyl)-D-lysine 98%
Nalpha-(tert-Butoxycarbonyl)-D-lysine
N^a-Boc-D-lysine, 98%
Purity: 98%
CAS No.: 106719-44-2
Molecular Formula: C11H22N2O4
Formula Weight: 246.30
Storage Temperature: 2°C to 8°C
Melting Point: 196-200°
Optical Rotation: -22°(c=2 in methanol)
MDL No.: MFCD00076956
Formula:
C₁₁H₂₂N₂O₄ MW: 246.31 g/mol Melting Pt: 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076956 CAS Number: 106719-44-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...