trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62957-03
H62957-06
AAH62957-03EA
219
USD
AAH62957-03
AAH62957-06
AAH62957-MD
trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde 97%
trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde
trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%
Purity: 97%
CAS No.: 412357-50-7
Molecular Formula: C15H19NO3
Formula Weight: 261.31
MDL No.: MFCD06657671
Formula:
C₁₅H₁₉NO₃ MW: 261.321 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06657671 CAS Number: 412357-50-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...