(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Glu-OH, N-Fmoc-L-glutamic Acid

H62936-06 H62936-14 H62936-22
AAH62936-06EA 46.2 USD
AAH62936-06 AAH62936-14 AAH62936-22
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid

N-Fmoc-L-glutamic acid, 95%


Purity: 95% CAS No.: 121343-82-6 Molecular Formula: C20H19NO6 Formula Weight: 369.37 Beilstein Reference: 3633624 MDL No.: MFCD00237657

Formula: C₂₀H₁₉NO₆
MW: 369.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237657
CAS Number: 121343-82-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR