trans-3-(Boc-amino)cyclohexanemethanol 97%

Supplier: Thermo Scientific Chemicals

H62934-03 H62934-MD
AAH62934-03EA 481 USD
AAH62934-03 AAH62934-MD
trans-3-(Boc-amino)cyclohexanemethanol 97%
trans-3-(Boc-amino)cyclohexanemethanol

trans-3-(Boc-amino)cyclohexanemethanol, 97%


Purity: 97% CAS No.: 920966-17-2 Molecular Formula: C12H23NO3 Formula Weight: 229.31 MDL No.: MFCD03844607

Formula: C₁₂H₂₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844607
CAS Number: 920966-17-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR