tert-Butyl (cis-4-aminocyclohexyl)carbamate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
cis-4-(Boc-amino)cyclohexylamine
H62925-06
H62925-14
AAH62925-03EA
70.72
USD
AAH62925-03
AAH62925-06
AAH62925-14
tert-Butyl (cis-4-aminocyclohexyl)carbamate 97%
tert-Butyl (cis-4-aminocyclohexyl)carbamate
cis-4-(Boc-amino)cyclohexylamine, 97%
Purity: 97%
CAS No.: 247570-24-7
Molecular Formula: C11H22N2O2
Formula Weight: 214.30
MDL No.: MFCD03844604
Formula:
C11H22N2O2 Storage Temperature: Ambient |
MDL Number:
MFCD03844604 CAS Number: 247570-24-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...