Trans-2-Benzyloxycarbonylaminocyclobutanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H62919-03 H62919-MD
AAH62919-03EA 662 USD
AAH62919-03 AAH62919-MD
Trans-2-Benzyloxycarbonylaminocyclobutanecarboxylic acid 97%
Trans-2-Benzyloxycarbonylaminocyclobutanecarboxylic acid

trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%


Purity: 97% CAS No.: 1212272-03-1 Molecular Formula: C13H15NO4 Formula Weight: 249.26 MDL No.: MFCD03844619

Formula: C₁₃H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD03844619
CAS Number: 1212272-03-1

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR