2-[cis-4-(Boc-amino)cyclohexyl]ethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

H62917-03 H62917-MD
AAH62917-03EA 175 USD
AAH62917-03 AAH62917-MD
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine 97%
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine

2-[cis-4-(Boc-amino)cyclohexyl]ethylamine, 97%


Purity: 97% CAS No.: 509143-12-8 Molecular Formula: C13H26N2O2 Formula Weight: 242.35 MDL No.: MFCD03844601

Formula: C₁₃H₂₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03844601
CAS Number: 509143-12-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR