DL-Glutamine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid

H62915-03 H62915-06
AAH62915-03EA 53.6 USD
AAH62915-03 AAH62915-06
DL-Glutamine 98%
DL-Glutamine

DL-Glutamine, 98%


Purity: 98% CAS No.: 585-21-7 Molecular Formula: C5H10N2O Formula Weight: 146.14 Melting Point: 177° MDL No.: MFCD00065103

Formula: C₅H₁₀N₂O₃
MW: 146.144 g/mol
Melting Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00065103
CAS Number: 6899-04-3
Merck Index: 14,4471

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR