trans-1-(Boc-amino)-4-ethynylcyclohexane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-Butyl trans-4-ethynylcyclohexylcarbamate

H62905-03 H62905-MD
AAH62905-03EA 382 USD
AAH62905-03 AAH62905-MD
trans-1-(Boc-amino)-4-ethynylcyclohexane 97%
trans-1-(Boc-amino)-4-ethynylcyclohexane

trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%


Purity: 97% CAS No.: 947141-86-8 Molecular Formula: C13H21NO2 Formula Weight: 223.31 MDL No.: MFCD12963985

Formula: C₁₃H₂₁NO₂
MW: 223.316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12963985
CAS Number: 947141-86-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR