cis-4-(Boc-amino)cyclohexanecarboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H62871-03 H62871-06 H62871-MD
AAH62871-03EA 166 USD
AAH62871-03 AAH62871-06 AAH62871-MD
cis-4-(Boc-amino)cyclohexanecarboxaldehyde 97%
cis-4-(Boc-amino)cyclohexanecarboxaldehyde

cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%


Purity: 97% CAS No.: 181308-56-5 Molecular Formula: C12H21NO3 Formula Weight: 227.3 MDL No.: MFCD03844599

Formula: C₁₂H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844599
CAS Number: 181308-56-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR