Fmoc-Lys(Z)-OH 98% (by HPLC)
Supplier: Thermo Scientific Chemicals
Synonyms:
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysine, Fmoc-Lys(Cbz)-OH, Fmoc-Lys(Z)-OH, Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, Nalpha-Fmoc-Nepsilon-Cbz-L-lysine
H62866-06
H62866-14
AAH62866-06EA
80.6
USD
AAH62866-06
AAH62866-14
Fmoc-Lys(Z)-OH 98% (by HPLC)
Fmoc-Lys(Z)-OH
N^a-Fmoc-N^e-benzyloxycarbonyl-L-lysine, 98%
Purity: 98%
CAS No.: 86060-82-4
Molecular Formula: C29H30N2O6
Formula Weight: 502.56
Storage Temperature: -30°C to -10°C
Melting Point: 110-120°
Beilstein Reference: 3578530
MDL No.: MFCD00065662
Formula:
C₂₉H₃₀N₂O₆ MW: 502.567 g/mol Melting Pt: 110…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065662 CAS Number: 86060-82-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...