Boc-S-trityl-D-cysteine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-S-trityl-D-cysteine, Boc-D-Cys(Trt)-OH
H62863-03
H62863-06
AAH62863-03EA
53.3
USD
AAH62863-03
AAH62863-06
Boc-S-trityl-D-cysteine 98%
Boc-S-trityl-D-cysteine
N-Boc-S-trityl-D-cysteine, 98%
Purity: 98%
CAS No.: 87494-13-1
Molecular Formula: C27H29NO4S
Formula Weight: 463.59
MDL No.: MFCD00236839
Formula:
C₂₇H₂₉NO₄S MW: 463.592 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236839 CAS Number: 87494-13-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...