N-Carbobenzoxyglycine methyl ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Cbz-glycine Methyl Ester, N-Benzyloxycarbonylglycine methyl ester, Z-Gly-OMe

H62858-06 H62858-14
AAH62858-06EA 49.2 USD
AAH62858-06 AAH62858-14 AAH62858-22
N-Carbobenzoxyglycine methyl ester 98%
N-Carbobenzoxyglycine methyl ester

N-Benzyloxycarbonylglycine methyl ester, 98%


Purity: 98% CAS No.: 1212-53-9 Molecular Formula: C11H13NO4 Formula Weight: 223.23 Melting Point: 22-26° Boiling Point: 147-151° MDL No.: MFCD00043631

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Boiling Pt: 147…151 °C
Melting Pt: 22…26 °C
Storage Temperature: Ambient
MDL Number: MFCD00043631
CAS Number: 1212-53-9
EINECS: 214-922-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR