Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Nδ-Boc-Nɑ-Fmoc-L-ornithine, Nδ-Boc-Nα-Fmoc-L-ornithine, Fmoc-Orn(Boc)-OH

H62814-06 H62814-14
AAH62814-03EA 29.69 USD
AAH62814-03 AAH62814-06 AAH62814-14
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine 96%
Ndelta-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine

N^d-Boc-N^a-Fmoc-L-ornithine, 96%


Purity: 96% CAS No.: 109425-55-0 Molecular Formula: C25H30N2O6 Formula Weight: 454.52 Storage Temperature: 2°C to 8°C Melting Point: 111-115° Beilstein Reference: 4772025 MDL No.: MFCD00065668

Formula: C₂₅H₃₀N₂O₆
MW: 454.52 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00065668
CAS Number: 109425-55-0

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR