Fmoc-Sar-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methylglycine, N-Fmoc-N-methylglycine, Fmoc-N-Me-Gly-OH
H62806-06
H62806-14
AAH62806-06EA
68
USD
AAH62806-06
AAH62806-14
Fmoc-Sar-OH 98%
Fmoc-Sar-OH
N-Fmoc-N-methylglycine, 98%
Purity: 98%
CAS No.: 77128-70-2
Molecular Formula: C18H17NO4
Formula Weight: 311.33
Storage Temperature: 2°C to 8°C
Beilstein Reference: 6660958
MDL No.: MFCD00151926
Formula:
C₁₈H₁₉NO₅ MW: 329.352 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151926 CAS Number: 77128-70-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...