trans-4-(Boc-aminomethyl)cyclohexanemethanol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-Butyl ((trans-4-(hydroxymethyl)cyclohexyl)methyl)carbamate

H62798-03 H62798-06
AAH62798-03EA 127 USD
AAH62798-03 AAH62798-06 AAH62798-MD
trans-4-(Boc-aminomethyl)cyclohexanemethanol 97%
trans-4-(Boc-aminomethyl)cyclohexanemethanol

trans-4-(Boc-aminomethyl)cyclohexanemethanol, 97%


Purity: 97% CAS No.: 172348-63-9 Molecular Formula: C13H25NO3 Formula Weight: 243.34 MDL No.: MFCD06657666

Formula: C₁₃H₂₅NO₃
MW: 243.347 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657666
CAS Number: 172348-63-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR